Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FUS Recombinant Rabbit Monoclonal Antibody (BLR023E)

Catalog No. PIMA544242
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544242 20 μL
PIMA544241 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA544242 Supplier Invitrogen™ Supplier No. MA544242
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE cell sections.

This gene encodes a multifunctional protein component of the heterogeneous nuclear ribonucleoprotein (hnRNP) complex. The hnRNP complex is involved in pre-mRNA splicing and the export of fully processed mRNA to the cytoplasm. This protein belongs to the FET family of RNA-binding proteins which have been implicated in cellular processes that include regulation of gene expression, maintenance of genomic integrity and mRNA/microRNA processing. Alternative splicing results in multiple transcript variants. Defects in this gene result in amyotrophic lateral sclerosis type 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FUS
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone BLR023E
Concentration 30 μg/mL
Conjugate Unconjugated
Formulation BBS with 0.1% BSA and 0.09% sodium azide; pH 8.2
Gene FUS
Gene Accession No. P35637, P56959
Gene Alias 16) in malignant liposarcoma); 16) malignant liposarcoma; 75 kDa DNA-pairing protein; ALS6; D430004D17Rik; D930039C12Rik; ETM4; FUS; FUS RNA binding protein; Fus1; fused in sarcoma; fused in sarcoma RNA binding protein; fusion (involved in t(12; fusion gene in myxoid liposarcoma; fusion, derived from t(12; fusion, derived from t(12;16) malignant liposarcoma; fus-like protein; heterogeneous nuclear ribonucleoprotein P2; hnRNP P2; HNRNPP2; oncogene FUS; Oncogene TLS; pigpen protein; POMP75; Protein pigpen; RNA-binding protein FUS; TLS; translocated in liposarcoma; translocated in liposarcoma protein
Gene Symbols FUS
Host Species Rabbit
Immunogen Peptide representing a region between residues 1 and 50 of human fusion (involved in t(12;16) in malignant liposarcoma).
Purification Method purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 233908, 2521
Target Species Human, Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.