Learn More
FUS Recombinant Rabbit Monoclonal Antibody (BLR023E), Bethyl Laboratories
Rabbit Recombit Monoclonal Antibody
$157.08 - $378.84
Specifications
Antigen | FUS |
---|---|
Clone | BLR023E |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot |
Classification | Recombinant Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1300
![]() |
Bethyl Laboratories, Inc
A700023T |
20 μL |
Each for $157.08
|
|
|||||
50-157-1299
![]() |
Bethyl Laboratories, Inc
A700023 |
100 μL |
Each for $378.84
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For ICC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE cell sections. For IHC, epitope retrieval with citrate buffer pH 6.0 is recommende
FUS/TLS was originally described as fused in Sarcoma is actually related to a number of different cellular processes and correlated to several diseases. FUS binds DNA as a single or double strand, and acts as a site for transcription factors to recognize. Since FUS is well-documented to be involved in liposarcoma, lipoma and leukaemia, it is believed that when its normal housekeeping functions related to transcription are disrupted, normal cells become cancerous. Interestingly, FUS is also associated with the fatal neuron disorder amyotrophic lateral sclerosis; interruption of its normal RNA-binding functions leads to incorrect cellular localization and leads to pathologies of the neuron. Thus, FUS regulation of cellular functions is a critical component of maintece of healthy cells.Specifications
FUS | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P35637, P56959 | |
233908, 2521 | |
IgG | |
Protein A | |
Antibody |
BLR023E | |
Recombinant Monoclonal | |
Liquid | |
RUO | |
BBS with 0.1% BSA and 0.09% sodium azide; pH 8.2 | |
ALS6, FUS1, HNRNPP2, POMP75, TLS, 75 kDa DNA-pairing protein, RNA-binding protein FUS, fus-like protein, fusion gene in myxoid liposarcoma, heterogeneous nuclear ribonucleoprotein P2, oncogene FUS, oncogene TLS, translocated in liposarcoma protein | |
Between 1 and 50 | |
Primary | |
4° C | |
FUS |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.