Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HYAL3 Monoclonal Antibody (3A3)
GREENER_CHOICE

Catalog No. PIMA524427
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524427 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524427 Supplier Invitrogen™ Supplier No. MA524427
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MTTQLGPALV LGVALCLGCG QPLPQVPERP FSVLWNVPSA HCEARFGVHL PLNSLGIIAN RGQHFHGQNM TIFYKNQLGL YPYFGPRGTA HNGGIPQALP LDRHLALAAY QIHHSLRPGF AGPAVLDWEE WCPLWAGNWG RRRAYQAASW AWAQQVFPDL DPQEQLYKAY TGFEQAARAL MEDTLRVAQA LRPHGLWGFY HYPACGNGWH SMASNYTGRC HAATLARNTQ LHWLWAASSA LFPSIYLPPR LPPAHHQAFV RHRLEEAFRV ALVGHRHPLP VLAYVRLTHR RSGRFLSQDD LVQSIGVSAA LGAAGVVLWG DLSLSSSEEE CWHLHDYLVD TLGPYVINVT RAAMACSHQR CHGHGRCARR DPGQMEAFLH LWPDGSLGDW KSFSCHCYWG WAGPTCQEPR PGPKEAV.

This gene encodes a member of the hyaluronidase family. Hyaluronidases are endoglycosidase enzymes that degrade hyaluronan, one of the major glycosaminoglycans of the extracellular matrix. The regulated turnover of hyaluronan plays a critical role in many biological processes including cell proliferation, migration and differentiation. The encoded protein may also play an important role in sperm function. This gene is one of several related genes in a region of chromosome 3p21.3 associated with tumor suppression, and the expression of specific transcript variants may be indicative of tumor status. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene, and some isoforms may lack hyaluronidase activity. This gene overlaps and is on the same strand as N-acetyltransferase 6 (GCN5-related), and some transcripts of each gene share a portion of the first exon.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HYAL3
Applications ELISA
Classification Monoclonal
Clone 3A3
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene HYAL3
Gene Accession No. O43820
Gene Alias Hyal3; hyal-3; Hyaluronidase; hyaluronidase 3; hyaluronidase-3; hyaluronoglucosaminidase 3; hyaluronoglucosaminidase-3; Hyl3; LUCA14; LUCA3; LuCa-3; Lung carcinoma protein 3; Minna14
Gene Symbols HYAL3
Host Species Mouse
Immunogen HYAL3 (AAH05896, 1 a.a. ~ 417 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8372
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.