Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fusidic acid sodium, Alfa Aesar™

Click to view available options
Quantity:
1g
5g
751-94-0
C31H47NaO6
538.701
MFCD09054714
HJHVQCXHVMGZNC-RDWMJMSASA-M
Fucidin
126963539
sodium;2-[(3R,4S,5S,8S,9R,10S,11R,13R,14S,16S)-16-acetyloxy-3,11-dihydroxy-4,8,10,14-tetramethyl-2,3,4,5,6,7,9,11,12,13,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-17-ylidene]-6-methylhept-5-enoate
CC1C2CCC3(C(C2(CCC1O)C)C(CC4C3(CC(C4=C(CCC=C(C)C)C(=O)[O-])OC(=O)C)C)O)C.[Na+]
751-94-0
C31H47NaO6
538.701
MFCD09054714
HJHVQCXHVMGZNC-RDWMJMSASA-M
Fucidin
126963539
sodium;2-[(3R,4S,5S,8S,9R,10S,11R,13R,14S,16S)-16-acetyloxy-3,11-dihydroxy-4,8,10,14-tetramethyl-2,3,4,5,6,7,9,11,12,13,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-17-ylidene]-6-methylhept-5-enoate
CC1C2CCC3(C(C2(CCC1O)C)C(CC4C3(CC(C4=C(CCC=C(C)C)C(=O)[O-])OC(=O)C)C)O)C.[Na+]
Merck Index 14,4317
Solubility Information Soluble in water
Formula Weight 538.69
Physical Form Powder
Color White
Quantity 1g
Sensitivity Moisture and Light Sensitive
Chemical Name or Material Fusidic acid sodium

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : LV5775000

storageNote1 : 2° to 8°C