Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fusion glycoprotein F0 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109300
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109300 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109300 Supplier Invitrogen™ Supplier No. PA5109300
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: Fusion glycoprotein F0 transfected HEK293 cell.

During virus entry, induces fusion of viral and cellular membranes leading to delivery of the nucleocapsid into the cytoplasm. The fusogenic activity is inactive untill entry into host cell endosome, where a furin-like protease cleaves off a small peptide between F1 and F2. Interacts directly with heparan sulfate and may participate in virus attachment. Furthermore, the F2 subunit was identifed as the major determinant of RSV host cell specificity. Later in infection, proteins F expressed at the plasma membrane of infected cells can mediate fusion with adjacent cells to form syncytia, a cytopathic effect that could lead to tissue necrosis. The fusion protein is also able to trigger p53-dependent apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fusion glycoprotein F0
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Host Species Rabbit
Immunogen Synthetic peptide within newcastle disease virus Fusion glycoprotein F0 aa 504-553.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.