Learn More
Cancer Diagnostics, Inc.™ Fusion Micro Scalpel
Turns microtome blades into a knife
$226.00 - $456.00
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
22-500-459
![]() |
Cancer Diagnostics, Inc.™
FUS000 |
Scalpel (Handle Only) |
Case of 50 for $226.00
|
|
|||||
22-500-460
![]() |
Cancer Diagnostics, Inc.™
FUS999 |
Scalpel Combo Pack |
Case of 50 for $456.00
|
|
|||||
Description
- Get more use out of your old microtome blades—simply secure blade into Fusion handle and use as a knife or scalpel
- Single-use handle and low-profile blades in dispenser pack available separately
- Combo pack includes 50 handles and 50 blades