Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FUSIP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562851
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562851 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562851 Supplier Thermo Scientific Supplier No. PA562851
Add to Cart
Add to Cart

FUSIP1 Polyclonal Antibody

FUSIP1 Polyclonal Antibody for ICC/IF, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen FUSIP1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SRSF10
Gene Accession No. O75494
Gene Alias 40 kDa SR-repressor protein; FUS interacting protein (serine/arginine-rich) 1; FUS interacting protein (serine-arginine rich) 1; Fus-associated protein with serine-arginine repeats; FUS-interacting protein (serine-arginine rich) 2; FUS-interacting serine-arginine-rich protein 1; Fusip1; FUSIP2; Neural-salient serine/arginine-rich protein; neural-salient SR protein; neural-specific SR protein; Nssr; NSSR1; NSSR2; PPP1R149; protein phosphatase 1, regulatory subunit 149; serine and arginine rich splicing factor 10; serine/arginine-rich splicing factor 10; serine/arginine-rich splicing factor 13A; serine-arginine repressor protein (40 kDa); SFRS13; SFRS13A; Splicing factor SRp38; splicing factor, arginine/serine-rich 13; splicing factor, arginine/serine-rich 13A; SR splicing factor 10; SRp38; SRrp40; Srsf10; Srsf13a; TASR; TASR1; TASR2; TLS-associated protein TASR; TLS-associated protein with Ser-Arg repeats; TLS-associated protein with SR repeats; TLS-associated serine-arginine protein; TLS-associated serine-arginine protein 1; TLS-associated serine-arginine protein 2; TLS-associated SR protein
Gene Symbols SRSF10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FUSIP1. Recombinant protein control fragment (Product #RP-104493).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10772
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.