Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FUSIP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541929
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541929 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541929 Supplier Invitrogen™ Supplier No. PA541929
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: TDSKTHYKSG SRYEKESRKK EPPRSKSQSR SQSRSRSKSR SRSWTSPKSS Sequence homology: Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

FUSIP1 is a member of the serine-arginine (SR) family of proteins, which is involved in constitutive and regulated RNA splicing. Members of this family are characterized by N-terminal RNP1 and RNP2 motifs, which are required for binding to RNA, and multiple C-terminal SR/RS repeats, which are important in mediating association with other cellular proteins. This protein can influence splice site selection of adenovirus E1A pre-mRNA. It interacts with the oncoprotein TLS, and abrogates the influence of TLS on E1A pre-mRNA splicing.This gene product is a member of the serine-arginine (SR) family of proteins, which is involved in constitutive and regulated RNA splicing. Members of this family are characterized by N-terminal RNP1 and RNP2 motifs, which are required for binding to RNA, and multiple C-terminal SR/RS repeats, which are important in mediating association with other cellular proteins. This protein can influence splice site selection of adenovirus E1A pre-mRNA. It interacts with the oncoprotein TLS, and abrogates the influence of TLS on E1A pre-mRNA splicing. Alternative splicing of this gene results in at least two transcript variants encoding different isoforms. In addition, transcript variants utilizing alternative polyA sites exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FUSIP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SRSF10
Gene Accession No. O75494
Gene Alias 40 kDa SR-repressor protein; FUS interacting protein (serine/arginine-rich) 1; FUS interacting protein (serine-arginine rich) 1; Fus-associated protein with serine-arginine repeats; FUS-interacting protein (serine-arginine rich) 2; FUS-interacting serine-arginine-rich protein 1; Fusip1; FUSIP2; Neural-salient serine/arginine-rich protein; neural-salient SR protein; neural-specific SR protein; Nssr; NSSR1; NSSR2; PPP1R149; protein phosphatase 1, regulatory subunit 149; serine and arginine rich splicing factor 10; serine/arginine-rich splicing factor 10; serine/arginine-rich splicing factor 13A; serine-arginine repressor protein (40 kDa); SFRS13; SFRS13A; Splicing factor SRp38; splicing factor, arginine/serine-rich 13; splicing factor, arginine/serine-rich 13A; SR splicing factor 10; SRp38; SRrp40; Srsf10; Srsf13a; TASR; TASR1; TASR2; TLS-associated protein TASR; TLS-associated protein with Ser-Arg repeats; TLS-associated protein with SR repeats; TLS-associated serine-arginine protein; TLS-associated serine-arginine protein 1; TLS-associated serine-arginine protein 2; TLS-associated SR protein
Gene Symbols SRSF10
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human FUSIP1 (aa 211-260).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10772
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.