Learn More
Invitrogen™ FUT1 Polyclonal Antibody

Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Kidney Tissue, SW620 whole cell, A549 whole cell. IHC: mouse intestine tissue, rat intestine tissue, human mammary cancer tissue.
Specifications
Specifications
| Antigen | FUT1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 500 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 4mg trehalose and 0.05mg sodium azide |
| Gene | FUT1 |
| Gene Accession No. | O09160, P19526, Q10980 |
| Gene Alias | 2-alpha-L-fucosyltransferase; alpha (1,2) fucosyltransferase; alpha 1,2-fucosyltransferase A; alpha 12-fucosyltransferase; alpha(1,2) fucosyltransferase 1; alpha(1,2)FT 1; alpha12-fucosyltransferase a; beta-galactoside alpha-1,2-fucosyltransferase; blood group H alpha 2-fucosyltransferase; Fta; Fucosyltransferase 1; fucosyltransferase 1 (galactoside 2-alpha-L-fucosyltransferase); fucosyltransferase 1 (galactoside 2-alpha-L-fucosyltransferase, H blood group); fucosyltransferase 1 (H blood group); FUT1; Futa; galactoside 2-alpha-L-fucosyltransferase 1; GDP-L-fucose:beta-D-galactoside 2-alpha-L-fucosyltransferase 1; H; H transferase; HH; HSC |
| Gene Symbols | FUT1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.