Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FUT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590570
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590570 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590570 Supplier Invitrogen™ Supplier No. PA590570
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VLAPEVDSRT PWRELQLHDW MSEEYADLRD PFLKLSGFPC SWTFFHHLRE QIRREFTLHD HLREEAQSVL GQLRLGRTGD RPRTFVGVHV RRGDYLQVMP QRWKGVVGDS AYLRQAMDWF RARHEAPVFV VTSNGMEWCK ENIDTSQGDV TFAGDGQEAT PWKDFALLTQ CNHTIMTIGT F; Positive Samples: Rat brain; Cellular Location: Golgi apparatus, Golgi stack membrane, Single-pass type II membrane protein.

FUT1 is a Golgi stack membrane protein that is involved in the creation of a precursor of the H antigen, which is required for the final step in the soluble A and B antigen synthesis pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FUT1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene FUT1
Gene Accession No. O09160, P19526, Q10980
Gene Alias 2-alpha-L-fucosyltransferase; alpha (1,2) fucosyltransferase; alpha 1,2-fucosyltransferase A; alpha 12-fucosyltransferase; alpha(1,2) fucosyltransferase 1; alpha(1,2)FT 1; alpha12-fucosyltransferase a; beta-galactoside alpha-1,2-fucosyltransferase; blood group H alpha 2-fucosyltransferase; Fta; Fucosyltransferase 1; fucosyltransferase 1 (galactoside 2-alpha-L-fucosyltransferase); fucosyltransferase 1 (galactoside 2-alpha-L-fucosyltransferase, H blood group); fucosyltransferase 1 (H blood group); FUT1; Futa; galactoside 2-alpha-L-fucosyltransferase 1; GDP-L-fucose:beta-D-galactoside 2-alpha-L-fucosyltransferase 1; H; H transferase; HH; HSC
Gene Symbols FUT1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 130-310 of human FUT1 (NP_0001391).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14343, 2523, 81919
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.