Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FUT10 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299776AF750
Description
FUT10 Polyclonal specifically detects FUT10 in Human samples. It is validated for ELISA.Specifications
FUT10 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
Alexa Fluor 750 | |
alpha 1,3-fucosyl transferase, alpha-(1,3)-fucosyltransferase 10, EC 2.4.1, EC 2.4.1.-, Fucosyltransferase 10, fucosyltransferase 10 (alpha (1,3) fucosyltransferase), Fucosyltransferase X, Fuc-TX, FucT-X, Galactoside 3-L-fucosyltransferase 10, MGC11141 | |
Produced in rabbits immunized with purified, recombinant FUT10 (Q6P4F1-1; Leu 32-Asp 479). | |
0.1 mL | |
Neuroscience | |
84750 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction