Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FUT10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562887
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562887 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562887 Supplier Invitrogen™ Supplier No. PA562887
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LGKFERKEFK SSSLQDGHTK MEEAPTHLNS FLKKEGLTFN RKRKWELDSY PIMLWWSPLT GETGRLGQCG ADACFFTINR TYLHHH Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 66%.

Probable fucosyltransferase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FUT10
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FUT10
Gene Accession No. Q6P4F1
Gene Alias alpha (1,3) fucosyltransferase; alpha 1,3-fucosyl transferase; alpha-(1,3)-fucosyltransferase 10; alpha3-fucosyltransferase; Fucosyltransferase 10; fucosyltransferase 10 (alpha (1,3) fucosyltransferase); fucosyltransferase X; FUCTX; fuc-TX; fucT-X; FUT10; fut10A; Galactoside 3-L-fucosyltransferase 10
Gene Symbols FUT10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FUT10. Recombinant protein control fragment (Product #RP-101490).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84750
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.