Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FUT10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109742
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109742 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109742 Supplier Invitrogen™ Supplier No. PA5109742
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NIDSLPLPRK AHHDWAVFHE ESPKNNYKLF HKPVITLFNY TATFSRHSHL PLTTQYLESI EVLKSLRYLV PLQSKNKLRK RLAPLVYVQS DCDPPSDRDS YVRELMTYIE VDSYGECLRN KDLPQQLKNP ASMDADGFYR IIAQYKFILA FENAVCDDYI TEKFWRPLKL GVVPVYYGSP SITDWLPSNK SAILVSEFSH PRELASYIRR LDSDDRLYEA YVEWKLKGEI SNQRLLTALR ERKWGVQDVN QDNYIDAFEC MVCTKVWANI RLQEKGLPPK RWEAEDTHLS CPEPTVFAFS PLRTPPLSSL REMWISSFEQ SKKEAQALRW LVDRNQNFSS QEFWGLVFKD.

Probable fucosyltransferase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FUT10
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.56 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene FUT10
Gene Accession No. Q5F2L2
Gene Alias alpha (1,3) fucosyltransferase; alpha 1,3-fucosyl transferase; alpha-(1,3)-fucosyltransferase 10; alpha3-fucosyltransferase; Fucosyltransferase 10; fucosyltransferase 10 (alpha (1,3) fucosyltransferase); fucosyltransferase X; FUCTX; fuc-TX; fucT-X; FUT10; fut10A; Galactoside 3-L-fucosyltransferase 10
Gene Symbols FUT10
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 130-479 of human FUT10 (NP_116053.3).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171167
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.