Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FUT11 Polyclonal Antibody
Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA548094
Description
In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) FUT3, rhFUT5, and rhFUT8 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
The FUT11 gene encodes for fructosyltransferase 11, an enzyme that is part of the glycosyltransferase family involved in the modification of glycoproteins and glycolipids by catalyzing the addition of fucose moieties. Enzymes like FUT11 play a critical role in the biosynthesis of Lewis antigen structures, significantly contributing to cell-cell adhesion, signal transduction, and immune recognition. Fucosylation, the process mediated by fructosyltransferases, is crucial for various biological functions, including the regulation of leukocyte trafficking and selectin binding, emphasizing its importance in immune responses and inflammation. While FUT11 is less characterized compared to other fucosyltransferases, altered expression patterns and functional abnormalities in these enzymes are linked to pathological conditions, such as cancer and inflammatory diseases, where changes in glycosylation can affect tumor progression, metastasis, and immune evasion. Ongoing research into FUT11 aims to elucidate its specific role in cellular processes and explore potential therapeutic applications by targeting fucosylation pathways to modify disease progression.
Specifications
FUT11 | |
Polyclonal | |
Unconjugated | |
FUT11 | |
3110009G03Rik; AI317183; alpha (1,3) fucosyltransferase; alpha-(1,3)-fucosyltransferase 11; alpha3-fucosyltransferase 11; EC 2.4.1; Fucosyltransferase 11; fucosyltransferase 11 (alpha (1,3) fucosyltransferase); fucosyltransferase XI; fuc-TXI; fucT-XI; Fut11; Galactoside 3-L-fucosyltransferase 11; MGC119338; MGC119339; MGC33202 | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
170384 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunoprecipitation, Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q495W5 | |
FUT11 | |
CHO-derived recombinant human Fucosyltransferase 11/FUT11 Gly25-Leu492. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction