Learn More
Invitrogen™ FUT11 Polyclonal Antibody

Description
Immunogen sequence: MKYLAYKQPG GITNQFLLDS LKHREWGVND PLLPNYLNGF ECFVCDYELA RLDAEKAHAA SPGDSPVFEP HIAQPSHMDC PVPTPGFGNV EEIPENDSWK EMWLQDYWQG LDQGEALTAM IHNNETEQTK FWDYL Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 89%.
Specifications
Specifications
| Antigen | FUT11 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | FUT11 |
| Gene Accession No. | Q495W5 |
| Gene Alias | 3110009G03Rik; AI317183; alpha (1,3) fucosyltransferase; alpha-(1,3)-fucosyltransferase 11; alpha3-fucosyltransferase 11; EC 2.4.1; Fucosyltransferase 11; fucosyltransferase 11 (alpha (1,3) fucosyltransferase); fucosyltransferase XI; fuc-TXI; fucT-XI; Fut11; Galactoside 3-L-fucosyltransferase 11; MGC119338; MGC119339; MGC33202 |
| Gene Symbols | FUT11 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.