Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FUT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553159
Description
Immunogen sequence: QRLAKIQAMW ELPVQIPVLA STSKALGPSQ LRGMWTINAI GRLGNQMGEY ATLYALAKMN GRPAFIPAQM HSTL Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 54%.
The protein encoded by this gene is a Golgi stack membrane protein that is involved in the creation of a precursor of the H antigen, which is required for the final step in the soluble A and B antigen synthesis pathway. This gene is one of two encoding the galactoside 2-L-fucosyltransferase enzyme. Two transcript variants encoding the same protein have been found for this gene.
Specifications
| FUT2 | |
| Polyclonal | |
| Unconjugated | |
| FUT2 | |
| 12FT 2; 12FT2; alpha (1,2) fucosyltransferase; alpha(1,2)FT 2; alpha(1,2)FT2; B12QTL1; Fucosyltransferase 2; fucosyltransferase 2 (secretor status included); FUT2; Galactoside 2-alpha-L-fucosyltransferase 2; GDP-L-fucose:beta-D-galactoside 2-alpha-L-fucosyltransferase 2; Se; SE2; SEC2; Secretor blood group alpha-2-fucosyltransferase; secretor factor; Secretory blood group protein 2; sej | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2524 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q10981 | |
| FUT2 | |
| Recombinant protein corresponding to Human FUT2. Recombinant protein control fragment (Product #RP-90487). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction