Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FUT2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5120940

This item is not returnable.
View return policy
Description
Positive test controls include: HT-29. The target is usually found in the following locations: Golgi apparatus, Golgi stack membrane, Single-pass type II membrane protein.
FUT2 Polyclonal antibody specifically detects FUT2 in Human, Mouse samples. It is validated for Western BlotSpecifications
FUT2 | |
Polyclonal | |
Unconjugated | |
FUT2 | |
12FT 2; 12FT2; alpha (1,2) fucosyltransferase; alpha(1,2)FT 2; alpha(1,2)FT2; B12QTL1; Fucosyltransferase 2; fucosyltransferase 2 (secretor status included); FUT2; Galactoside 2-alpha-L-fucosyltransferase 2; GDP-L-fucose:beta-D-galactoside 2-alpha-L-fucosyltransferase 2; Se; SE2; SEC2; Secretor blood group alpha-2-fucosyltransferase; secretor factor; Secretory blood group protein 2; sej | |
Recombinant fusion protein containing a sequence corresponding to amino acids 29-150 of human FUT2 (NP_001091107.1). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
0.54 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q10981, Q9JL27 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14344, 2524 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title