Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FUT3 Mouse anti-Human, Unconjugated, Clone: 2-25LE, Abnova™

Catalog No. 89337690 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-690 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-690 Supplier Abnova Corporation Supplier No. MAB21197
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

FUT3 Monoclonal antibody specifically detects FUT3 in Human samples. It is validated for IHC-P, IF.

Specifications

Antigen FUT3
Applications Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone 2-25LE
Conjugate Unconjugated
Description fucosyltransferase 3 (galactoside 3(4)-L-fucosyltransferase, Lewis blood group)
Gene FUT3
Gene Alias CD174, FT3B, FucT-III, LE, Les, MGC131739
Host Species Mouse
Immunogen Mucin isolated from a human ovarian cyst fluid
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 2525
Target Species Human
Content And Storage Store at 4°C
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.