Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FUT3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111250
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111250 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111250 Supplier Invitrogen™ Supplier No. PA5111250
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KDLARYLQEL DKDHARYLSY FRWRETLRPR SFSWAL Highest antigen sequence indentity to the following orthologs: Mouse - 39%, Rat - 39%.

FUT3 comprises a set of fucosylated glycosphingolipids that are synthesized by exocrine epithelial cells and circulate in body fluids. The glycosphingolipids function in embryogenesis, tissue differentiation, tumor metastasis, inflammation, and bacterial adhesion. They are secondarily absorbed to red blood cells giving rise to their Lewis phenotype. This protein is a member of the fucosyltransferase family, which catalyzes the addition of fucose to precursor polysaccharides in the last step of Lewis antigen biosynthesis. It encodes an enzyme with alpha(1,3)-fucosyltransferase and alpha(1,4)-fucosyltransferase activities.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FUT3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FUT3
Gene Accession No. P21217
Gene Alias 4-fucosyltransferase; alpha-(1,3/1,4)-fucosyltransferase; blood group Lewis alpha-4-fucosyltransferase; CD174; FT3B; Fucosyltransferase 3; fucosyltransferase 3 (galactoside 3(4)-L-fucosyltransferase, Lewis blood group); fucosyltransferase 3 (Lewis blood group); fucosyltransferase III; FucT-III; Fuc-TV; FucT-V; FUT3; Galactoside 3(4)-L-fucosyltransferase; LE; Les; Lewis FT; Lewis Y
Gene Symbols FUT3
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human FUT3. Recombinant protein control fragment (Product #RP-104414).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2525
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.