Learn More
FUT4 Monoclonal Antibody (476CT26.8.4)

Description
FUT4 Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | FUT4 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 476CT26.8.4 |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | FUT4 |
| Gene Accession No. | P22083 |
| Gene Alias | 3-fucosyl-N-acetyl-lactosamine epitope; AI451562; alpha (1,3) fucosyltransferase, myeloid specific; alpha (1,3) fucosyltransferase, myeloid-specific; alpha 1,3-fucosyltransferase; alpha 1,3-fucosyltransferase Fuc-T (similar to mouse Fut4); alpha 13-fucosyltransferase Fuc-T (similar to mouse Fut4); alpha-(1,3)-fucosyltransferase 4; CD15; ELAM ligand fucosyltransferase; ELAM-1 ligand fucosyltransferase; ELFT; FAL; FCT3A; fucosyltransferase 4; fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific); fucosyltransferase IV; Fuct; FUC-TIV; FucT-IV; FUT4; FUTIV; Galactoside 3-L-fucosyltransferase; Lewis x; LeX; Ssea1; SSEA-1; stage-specific embryonic antigen 1 |
| Gene Symbols | FUT4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.