Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FUT4 Mouse anti-Human, Unconjugated, Clone: FUT4/815 + BRA-4F1, Abnova™

Catalog No. 89337492 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-492 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-492 Supplier Abnova Corporation Supplier No. MAB20999
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

FUT4 Monoclonal antibody specifically detects FUT4 in Human samples. It is validated for IHC-P, IF, Flow Cyt.

Specifications

Antigen FUT4
Applications Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone FUT4/815 + BRA-4F1
Conjugate Unconjugated
Description fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific)
Gene FUT4
Gene Alias CD15, ELFT, FCT3A, FUC-TIV, FUTIV
Host Species Mouse
Immunogen Purified human neutrophils (FUT4/815); Myelomonocytic leukemia cells (BRA-4F1)
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 2526
Target Species Human
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.