Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FUT4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

Supplier:  Proteintech Group Inc 194971AP150UL

Encompass_Preferred

 View more versions of this product

Catalog No. 50-173-2417


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

19497-1-AP antibody detects the native band around 59 kDa and glycosylated isoform proteins around 95-140 kDa in SDS-PAGE.

FUT4 (Fucosyltransferase 4) is a Protein Coding gene. Diseases associated with FUT4 include Colon Adenocarcinoma and Liver Lymphoma. Among its related pathways are Glycosphingolipid biosynthesis - lacto and neolacto series and Mannose type O-glycan biosynthesis. Gene Ontology (GO) annotations related to this gene include fucosyltransferase activity and alpha-(1->3)-fucosyltransferase activity. The product of this gene transfers fucose to N-acetyllactosamine polysaccharides to generate fucosylated carbohydrate structures. It catalyzes the synthesis of the non-sialylated antigen, Lewis x (CD15).
Specifications

Specifications

FUT4
Polyclonal
Unconjugated
FUT4
Alpha (1,3) fucosyltransferase, ELFT, FCT3A, FUC TIV, Fucosyltransferase 4, Fucosyltransferase IV, FucT IV, FUT4, FUTIV
Rabbit
Antigen Affinity Chromatography
RUO
14345, 2526
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P22083, Q11127
Fut4
Peptide
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.