Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FUT6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565366
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565366 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565366 Supplier Invitrogen™ Supplier No. PA565366
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RVSQDDPTVYP NGSRFPDSTG T Highest antigen sequence identity to the following orthologs - mouse 45%, rat 45%.

FUT6 is a Golgi stack membrane protein that is involved in the creation of sialyl-Lewis X, an E-selectin ligand.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FUT6
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FUT6
Gene Accession No. P51993
Gene Alias alpha-(1,3)-fucosyltransferase 6; FCT3A; FT1A; Fucosyltransferase 6; fucosyltransferase 6 (alpha (1,3) fucosyltransferase); fucosyltransferase VI; Fuc-TVI; FucT-VI; FUT6; Galactoside 3-L-fucosyltransferase
Gene Symbols FUT6
Host Species Rabbit
Immunogen Recombinant Human FUT6. Recombinant protein control fragment (Product #RP-106050).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2528
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.