Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FUT6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596261
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596261 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596261 Supplier Invitrogen™ Supplier No. PA596261
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WLEPWSGQPA HPPLNLSAKT ELVAWAVSNW GPNSARVRYY QSLQAHLKVD VYGRSHKPLP QGTMMETLSR YKFYLAFENS LHPDYITEKL WRNALEAWAV PVVLGPSRSN YERFLPPDAF IHVDDFQSPK DLARYLQELD KDHARYLSYF RWRETLRPRS FSWALAFCKA CWKLQEESRY QTRGIAAWFT; Positive Samples: SW480; Cellular Location: Golgi apparatus, Golgi stack membrane, Single-pass type II membrane protein.

FUT6 is a Golgi stack membrane protein that is involved in the creation of sialyl-Lewis X, an E-selectin ligand.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FUT6
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FUT6
Gene Accession No. P51993
Gene Alias alpha-(1,3)-fucosyltransferase 6; FCT3A; FT1A; Fucosyltransferase 6; fucosyltransferase 6 (alpha (1,3) fucosyltransferase); fucosyltransferase VI; Fuc-TVI; FucT-VI; FUT6; Galactoside 3-L-fucosyltransferase
Gene Symbols FUT6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 170-359 of human FUT6 (NP_0001411).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2528
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.