Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FUT6 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30477820UL
Description
FUT6 Polyclonal specifically detects FUT6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FUT6 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
2528 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry | |
Unconjugated | |
alpha-(1,3)-fucosyltransferase, EC 2.4.1, FCT3AFuc-TVI, FLJ40754, FT1A, Fucosyltransferase 6, fucosyltransferase 6 (alpha (1,3) fucosyltransferase), Fucosyltransferase VI, FucT-VIEC 2.4.1.65, Galactoside 3-L-fucosyltransferase | |
Recombinant fusion protein containing a sequence corresponding to amino acids 170-359 of human FUT6 (NP_000141.1). | |
20 μL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction