Learn More
Invitrogen™ FUT8 Polyclonal Antibody

Description
Immunogen sequence: MTDLYYLSQTD GAGDWREKEA KDLTELVQRR ITYLQNPKDC SKAKKLVCNI NKGCGYGCQL HHVVYCFMIA YGTQRTLILE SQNWRYATGG WETVFRP Highest antigen sequence identity to the following orthologs - mouse 99%, rat 99%.
Specifications
Specifications
| Antigen | FUT8 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | FUT8 |
| Gene Accession No. | Q9BYC5 |
| Gene Alias | alpha (1,6) fucosyltransferase; alpha-(1,6)-fucosyltransferase; alpha-1,6-fucosyltransferase; alpha1-6FucT; EC 2.4.1.68; Fucosyltransferase 8; fucosyltransferase 8 (alpha (1,6) fucosyltransferase); FUT8; GDP-fucose--glycoprotein fucosyltransferase; GDP-L-Fuc:N-acetyl-beta-D-glucosaminide alpha1,6-fucosyltransferase; Glycoprotein 6-alpha-L-fucosyltransferase; MGC26465 |
| Gene Symbols | FUT8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.