Learn More
Merit Medical Futura™ Safety Scalpel
Click to view available options
Size:
# 10
# 11
# 15
Description
Retractable blades; color-coded.
- Clinician controls the in-use or hidden position of the blade via a locking inset
- Completely disposable, the scalpel's compact design conserves disposal container space
Order Info
Blades are precision sharpened and available in multiple sizes to accommodate various applications.
Specifications
Specifications
| Product Type | Safety Scalpel |
| For Use With (Application) | To Protect Healthcare Workers from Accidental Cuts associated with Traditional Scalpels |
| Size | # 11 |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Handle Material | Polystyrene |