Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FUZ Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559724
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559724 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559724 Supplier Invitrogen™ Supplier No. PA559724
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLRNFYTLVT STHFPPEPGP PEKTEDEVYQ AQLPRACYLV LGTEEPGTGV RLVALQLGLR RLLLLLSPQS PTHGLRSLAT HTLHALTP Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 78%.

FUZ is the human homolog of Drosophila fy protein and belongs to the fuzzy family. The fy gene of Drosophila encodes a novel four-pass transmembrane protein that plays two roles in the development of hairs on the Drosophila wing. The first is to specify the correct orientation of the hair by restricting its initiation to the distal vertex of the cell. The second role is to permit the development of just a single cell hair by maintaining the integrity of the F-actin and microtubule arrays that are required for hair development. In humans, the protein is involved in cytoskeleton function and cell polarity in epithelial cells. The human fuz gene is localized on chromosome 19q13.33.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FUZ
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FUZ
Gene Accession No. Q9BT04
Gene Alias 2600013E07Rik; b2b1273Clo; Fuz; fuzzy homolog; fuzzy homolog (Drosophila); fuzzy planar cell polarity protein; FY; NTD; Protein fuzzy homolog; RGD1310608
Gene Symbols FUZ
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FUZ. Recombinant protein control fragment (Product #RP-98850).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80199
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.