Learn More
Invitrogen™ FUZ Polyclonal Antibody

Description
FUZ Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | FUZ |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA and 0.05% sodium azide |
| Gene | FUZ |
| Gene Accession No. | Q3B756, Q3UYI6, Q9BT04 |
| Gene Alias | 2600013E07Rik; b2b1273Clo; Fuz; fuzzy homolog; fuzzy homolog (Drosophila); fuzzy planar cell polarity protein; FY; NTD; Protein fuzzy homolog; RGD1310608 |
| Gene Symbols | FUZ |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.