Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FVT1 Monoclonal Antibody (2B2-3C11)
GREENER_CHOICE

Catalog No. PIMA518923
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518923 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518923 Supplier Invitrogen™ Supplier No. MA518923
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: FVLLLYMVSP LISPKPLALP GAHVVVTGGS SGIGKCIAIE CYKQGAFITL VARNEDKLLQ AKKEIEMHSI NDKQVVLCIS VDVSQDYNQV ENVIKQAQEK LGPVDMLVNC AGMAVSGKFE DLEVSTFERL MSINYLGSVY PSRAVITTMK ERRVGRIVFV SSQAGQLGLF GFTAYSASKF AIRGLAEALQ MEVKPYNVYI TVAYPPDTDT PGFAEENRTK PLETRLISET TSVCKPEQVA KQIVKDAIQG NFNSSLGSDG YMLSALTCGM APVTSITEGL QQVVTMGLFR TIALFYLGSF DSIVRRCMMQ REKSENADKT A.

The protein encoded by this gene catalyzes the reduction of 3-ketodihydrosphingosine to dihydrosphingosine. The putative active site residues of the encoded protein are found on the cytosolic side of the endoplasmic reticulum membrane. A chromosomal rearrangement involving this gene is a cause of follicular lymphoma, also known as type II chronic lymphatic leukemia. The mutation of a conserved residue in the bovine ortholog causes spinal muscular atrophy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FVT1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2B2-3C11
Concentration 0.35 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Kdsr
Gene Accession No. Q06136
Gene Alias 3-dehydrosphinganine reductase; 3-ketodihydrosphingosine reductase; 3-ketodihydrosphingosine reductase; LOW QUALITY PROTEIN: 3-ketodihydrosphingosine reductase; 6330410P18Rik; 9430079B08Rik; DHSR; follicular lymphoma variant translocation 1; follicular variant translocation protein 1; Follicular variant translocation protein 1 homolog; FVT1; FVT-1; KDS reductase; kdsr; MGC137214 protein; SDR35C1; Short chain dehydrogenase/reductase family 35C member 1; short chain dehydrogenase/reductase family 35C, member 1; unm_muz106; zgc:55947
Gene Symbols Kdsr
Host Species Mouse
Immunogen KDSR (AAH08797.1, 12 a.a. ~ 332 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2531
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.