Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FVT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531334
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531334 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531334 Supplier Invitrogen™ Supplier No. PA531334
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (92%), Rat (92%), Xenopus laevis (81%), Chicken (84%), Rhesus Monkey (99%), Chimpanzee (100%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene catalyzes the reduction of 3-ketodihydrosphingosine to dihydrosphingosine. The putative active site residues of the encoded protein are found on the cytosolic side of the endoplasmic reticulum membrane. A chromosomal rearrangement involving this gene is a cause of follicular lymphoma, also known as type II chronic lymphatic leukemia. The mutation of a conserved residue in the bovine ortholog causes spinal muscular atrophy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FVT1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Kdsr
Gene Accession No. Q06136
Gene Alias 3-dehydrosphinganine reductase; 3-ketodihydrosphingosine reductase; 3-ketodihydrosphingosine reductase; LOW QUALITY PROTEIN: 3-ketodihydrosphingosine reductase; 6330410P18Rik; 9430079B08Rik; DHSR; follicular lymphoma variant translocation 1; follicular variant translocation protein 1; Follicular variant translocation protein 1 homolog; FVT1; FVT-1; KDS reductase; kdsr; MGC137214 protein; SDR35C1; Short chain dehydrogenase/reductase family 35C member 1; short chain dehydrogenase/reductase family 35C, member 1; unm_muz106; zgc:55947
Gene Symbols Kdsr
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human FVT1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2531
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.