Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fx1A Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982855
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2855 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2855 Supplier Bioss Supplier No. BS1338R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Binding of anti-Fx1A to Heymann nephritis antigens (HA) on rat glomerular epithelial cells (GECs) in culture leads to capping and disappearance of antigens from the cell surface. This process may contribute to the formation of glomerular subepithelial immune deposits in vivo. The authors differentially extracted GECs to determine whether HA redistribution is mediated by cytoskeletal components. Observations were made by phase-contrast and immunofluorescence microscopy on primary and passaged GECs in monolayer culture and by spectrofluorimetry on GECs in suspension.

Specifications

Antigen Fx1A
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CD59
Gene Accession No. P13987
Gene Symbols CD59
Host Species Rabbit
Immunogen Fx1A protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 966
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.