Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FXN Monoclonal Antibody (OTI3C3), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA504352S


Description
FXN Monoclonal antibody specifically detects FXN in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
FXN | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P20810 | |
Mouse | |
Affinity chromatography | |
RUO | |
831 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Immunocytochemistry, Western Blot | |
OTI3C3 | |
Unconjugated | |
Cast | |
CyaY; d-FXN; DKEYP-118A9.1; FA; FARR; frataxin; Frataxin intermediate form; Frataxin mature form; Frataxin(56-210); Frataxin(78-210); Frataxin(81-210); Frataxin, mitochondrial; FRDA; Friedreich ataxia; Friedreich ataxia protein; FXN; i-FXN; m56-FXN; m78-FXN; m81-FXN; RGD1565754; X25; zgc:158218 | |
Full length human recombinant protein of human CAST produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction