Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FXN Mouse anti-Human, Unconjugated, Clone: FXN/2124, Abnova™

Catalog No. 89337485 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-485 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-485 Supplier Abnova Corporation Supplier No. MAB20992
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

FXN Monoclonal antibody specifically detects FXN in Human samples. It is validated for IHC-P.

Specifications

Antigen FXN
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone FXN/2124
Conjugate Unconjugated
Description frataxin
Gene FXN
Gene Alias CyaY, FA, FARR, FRDA, MGC57199, X25
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 57-210 of human FXN
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 2395
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.