Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FXR/NR1H4 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB400153PE
Description
FXR/NR1H4 Polyclonal antibody specifically detects FXR/NR1H4 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
FXR/NR1H4 | |
Polyclonal | |
Western Blot, Immunocytochemistry/ Immunofluorescence | |
bile acid receptor, farnesoid X receptor, Farnesoid X-activated receptor, Farnesol receptor HRR-1, FXRBAR, HRR-1, HRR1MGC163445, Nuclear receptor subfamily 1 group H member 4, nuclear receptor subfamily 1, group H, member 4, Retinoid X receptor-interacting protein 14, RIP14RXR-interacting protein 14 | |
A synthetic peptide made to the C-terminus of the human FXR protein. [UniProt# Q96RI1] | |
0.1 mL | |
Cancer, Chromatin Research, Lipid and Metabolism, Signal Transduction | |
9971 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunocytochemistry | |
PE | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction