Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FXR/NR1H4 Antibody (OTI4F12), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP27126A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27126A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27126A94 Supplier Novus Biologicals Supplier No. NBP271526AF594
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FXR/NR1H4 Monoclonal specifically detects FXR/NR1H4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen FXR/NR1H4
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4F12
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias bile acid receptor, farnesoid X receptor, Farnesoid X-activated receptor, Farnesol receptor HRR-1, FXRBAR, HRR-1, HRR1MGC163445, Nuclear receptor subfamily 1 group H member 4, nuclear receptor subfamily 1, group H, member 4, Retinoid X receptor-interacting protein 14, RIP14RXR-interacting protein 14
Gene Symbols NR1H4
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 21-297 of human FXR/NR1H4 (NP_005114) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 9971
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.