Learn More
Invitrogen™ FXR Recombinant Rabbit Monoclonal Antibody (4Z4G5)
Rabbit Recombinant Monoclonal Antibody
$169.00 - $419.00
Specifications
Antigen | FXR |
---|---|
Clone | 4Z4G5 |
Concentration | 1.63 mg/mL |
Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
Applications | ELISA, Western Blot |
Description
Immunogen sequence: MVMQFQGLEN PIQISPHCSC TPSGFFMEMM SMKPAKGVLT EQVAGPLGQN LEVEPYSQYS NVQFPQVQPQ ISSSSYYSNL GFYPQQPEEW YSPGIYELRR MPAETLYQGE TEVAEMPVTK KPRMGASAGR IKGDELCVVC GDRASGYHYN ALTCEGCKGF FRRSITKNAV YKCKNGGNCV MDMYMRRKCQ ECRLRKCKEM GMLAECMYTG LLTEIQCKSK RLRKNVKQHA.
FXR (farnesoid X receptor; NR1H4) belongs to nuclear receptor superfamily of ligand-activated transcription factors (NR1 subfamily) and acts as a receptor for bile acids such as chenodeoxycholic acid, lithocholic acid and deoxycholic acid. FXR plays an essential role in bile acid homeostasis through the regulation of genes involved in bile acid synthesis, conjugation and enterohepatic circulation. FXR also regulates lipid and glucose homeostasis and is involved innate immune response. The FXR-RXR heterodimer binds predominantly to farnesoid X receptor response elements (FXREs) containing two inverted repeats of the consensus sequence 5'-AGGTCA-3' in which the monomers are spaced by 1 nucleotide (IR-1) but also to tandem repeat DR1 sites with lower affinity, and can be activated by either FXR or RXR-specific ligands. In the liver FXR activates transcription of the corepressor NR0B2 thereby indirectly inhibiting CYP7A1 and CYP8B1 (involved in BA synthesis) implicating at least in part histone demethylase KDM1A resulting in epigenomic repression, and SLC10A1/NTCP (involved in hepatic uptake of conjugated BAs). FXR activates transcription of SLC27A5/BACS and BAAT (involved in BA conjugation), ABCB11/BSEP (involved in bile salt export) by directly recruiting histone methyltransferase CARM1, and ABCC2/MRP2 (involved in secretion of conjugated BAs) and ABCB4 (involved in secretion of phosphatidylcholine in the small intestine). (From Uniprot)Specifications
FXR | |
1.63 mg/mL | |
ELISA, Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
Q60641, Q62735, Q96RI1 | |
20186, 60351, 9971 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-230 of human FXR/NR1H4 (NP_001193922.1). | |
Antibody |
4Z4G5 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Recombinant Monoclonal | |
Liquid | |
IgG | |
Human, Mouse, Rat | |
NR1H4 | |
AI957360; BAR; bile acid receptor; farnesoid X activated receptor; farnesoid X nuclear receptor; farnesoid X-activated receptor; Farnesol receptor HRR-1; FXR; HRR1; HRR-1; hypothetical protein LOC540528; MGC163445; NR1H4; Nuclear receptor subfamily 1 group H member 4; nuclear receptor subfamily 1, group H, member 4; retinoid X receptor-interacting protein 14; RIP14; RXR-interacting protein 14; Rxrip14 | |
NR1H4 | |
Primary | |
Protein A |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.