Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FXR1 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297956APC
Description
FXR1 Polyclonal antibody specifically detects FXR1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
FXR1 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
fragile X mental retardation syndrome-related protein 1, fragile X mental retardation, autosomal homolog 1, FXR1P, hFXR1p | |
Produced in rabbits immunized with E. coli-derived Human FXR1 fragment. | |
0.1 mL | |
Apoptosis, Immune System Diseases, Immunology | |
8087 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction