Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FXR1 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298797G
Description
FXR1 Polyclonal specifically detects FXR1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
FXR1 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
fragile X mental retardation syndrome-related protein 1, fragile X mental retardation, autosomal homolog 1, FXR1P, hFXR1p | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human FXR1. | |
0.1 mL | |
Apoptosis, Immune System Diseases, Immunology | |
8087 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction