Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FXR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541734
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541734 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541734 Supplier Invitrogen™ Supplier No. PA541734
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: EQLRQIGSRS YSGRGRGRRG PNYTSGYGTN SELSNPSETE SERKDELSDW Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

FXR1 is an RNA binding protein that interacts with the functionally-similar proteins FMR1 and FXR2. It may regulate intracellular transport and local transltion of certain mRNAs. These proteins shuttle between the nucleus and cytoplasm and associate with polyribosomes, predominantly with the 60S ribosomal subunit. FXR1 is required for embryonic and postnatal development of muscle tissue. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FXR1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Fxr1
Gene Accession No. P51114
Gene Alias 1110050J02Rik; 9530073J07Rik; AI851072; FMR1 autosomal homolog 1; fragile X mental retardation autosomal homolog variant p1K; fragile X mental retardation autosomal homolog variant p2K; fragile X mental retardation autosomal homolog variant p4K; fragile X mental retardation autosomal homolog variant p5FK; fragile X mental retardation gene 1, autosomal homolog; fragile X mental retardation gene, autosomal homolog; fragile X mental retardation syndrome-related protein 1; fragile X mental retardation, autosomal homolog 1; fragile X mental retardation-related protein 1; fragile-X-related protein 1; Fxr1; Fxr1h; Fxr1p; hFXR1p; hypothetical protein LOC536793; mFxr1p
Gene Symbols Fxr1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human FXR1 (aa 373-422).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8087
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.