Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FXR2 Antibody (1G2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
$206.50 - $499.50
Specifications
Antigen | FXR2 |
---|---|
Clone | 1G2 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:500-1:2500, Immunohistochemistry 20 ug/ml, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 20 ug/ml |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB300259SS
![]() |
Novus Biologicals
NB300259SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB300259
![]() |
Novus Biologicals
NB300259 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
FXR2 Monoclonal specifically detects FXR2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
FXR2 | |
1.0 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cellular Markers, Chromatin Research, Neurodegeneration, Neuroscience | |
P51116 | |
9513 | |
Recombinant C-terminal fragment of human FXR2 (residues 414-658). [UniProt# P51116] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1G2 | |
Western Blot 1:500-1:2500, Immunohistochemistry 20 ug/ml, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 20 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
FMR1L2, fragile X mental retardation syndrome-related protein 2, fragile X mental retardation, autosomal homolog 2, fragile X-mental retardation 1-like 2, FXR2P | |
FXR2 | |
IgG1 κ | |
Protein G purified | |
80 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title