Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FXR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109743
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109743 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109743 Supplier Invitrogen™ Supplier No. PA5109743
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RRRTDEDRTV MDGGLESDGP NMTENGLEDE SRPQRRNRSR RRRNRGNRTD GSISGDRQPV TVADYISRAE SQSRQRPPLE RTKPSEDSLS GQKGDSVSKL P.

Fragile X syndrome is caused by the absence of the fragile X mental-retardation protein (FMRP). FMRP is the archetype of a class of cytoplasmic mRNA-binding proteins that includes the fragile X-related 1 and 2 proteins (FXR1 and FXR2). The fragile X-related proteins FXR1 and FXR2 contain a functional nucleolar-targeting signal equivalent to the HIV-1 regulatory proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FXR2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.07 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FXR2
Gene Accession No. P51116, Q9WVR4
Gene Alias FMR1 autosomal homolog 2; FMR1L2; fragile X mental retardation gene 2, autosomal homolog; fragile X mental retardation syndrome-related protein 2; fragile X mental retardation, autosomal homolog 2; fragile X mental retardation-related protein 2; fragile X-mental retardation 1-like 2; Fxr2; Fxr2h; FXR2P
Gene Symbols FXR2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 550-650 of human FXR2 (NP_004851.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23879, 9513
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.