Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FXR2 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | FXR2 |
---|---|
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
FXR2 Polyclonal specifically detects FXR2 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
FXR2 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
FMR1L2, fragile X mental retardation syndrome-related protein 2, fragile X mental retardation, autosomal homolog 2, fragile X-mental retardation 1-like 2, FXR2P | |
A synthetic peptide corresponding to a sequence within amino acids 550-650 of human FXR2 (NP_004851.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cellular Markers, Chromatin Research, Neurodegeneration, Neuroscience | |
PBS with 50% glycerol, pH7.3. | |
9513 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title