Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FXYD2-FXYD6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584978
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584978 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584978 Supplier Invitrogen™ Supplier No. PA584978
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LRPQEMRKPR WRTSSPPMQQ SPRKQR Highest antigen sequence indentity to the following orthologs: Mouse - 46%, Rat - 46%.

This locus represents naturally occurring read-through transcription between the neighboring FXYD domain-containing ion transport regulator 6 (GeneID 53826) and sodium/potassium-transporting ATPase subunit gamma (GeneID 486) genes on chromosome 11. One read-through transcript produces a fusion protein that shares sequence identity with each individual gene product, while another read-through transcript encodes a protein that has a distinct C-terminus and only shares sequence identity with the upstream locus (GeneID 53826). [provided by RefSeq, Aug 2011].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FXYD2-FXYD6
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FXYD6-FXYD2
Gene Accession No. 0
Gene Alias FXYD6-FXYD2; FXYD6-FXYD2 readthrough; FXYD6-FXYD2 read-through protein
Gene Symbols FXYD6-FXYD2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FXYD6-FXYD2. Recombinant protein control fragment (Product #RP-108650).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100533181
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.