Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FXYD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104165
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104165 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104165 Supplier Invitrogen™ Supplier No. PA5104165
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total FXYD2.

The human FXYD2 (pronounced fix-id) gene maps to chromosome 11q23 and encodes the gamma subunit of Na/K-ATPase (NKA). FXYD2 gamma subunit modulates NKA activity by inducing ion channel activity. The mammalian FXYD family maintains Na+ and K+ gradients between the intracellular and extracellular milieus of cells in processes such as renal Na+-reabsorption, muscle contraction and neuronal excitability. FXYDs are single-span membrane proteins that share a 35 amino acid signature domain, beginning with the sequence PFXYD and containing 7 invariant and 6 conserved amino acids. Other members of the FXYD family include FXYD1 (PLM, phospholemman), FXYD3 (Mat8, mammary tumor protein 8), FXYD4 (CHIF) and FXYD5 (RIC).
TRUSTED_SUSTAINABILITY

Specifications

Antigen FXYD2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Fxyd2
Gene Accession No. P54710, Q04646
Gene Alias ATP1C; Atp1g1; ATPase, Na+/K+ transporting, gamma 1 polypeptide; FXTD domain-containing ion transport regulator 2; FXYD domain containing ion transport regulator 2; FXYD domain-containing ion transport regulator 2; FXYD2; gamma subunit of sodium potassium ATPase; GNAKATP; HOMG2; Na(+)/K(+) ATPase subunit gamma; regulator 2; Sodium Potassium ATPase; sodium pump gamma chain; sodium/potassium-transporting ATPase gamma chain; sodium/potassium-transporting ATPase subunit gamma
Gene Symbols Fxyd2
Host Species Rabbit
Immunogen A synthesized peptide derived from human FXYD2(Accession P54710), corresponding to amino acid residues I32-P66.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11936, 486
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.