Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FXYD2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566670
Description
Immunogen sequence: MTGLSMDGGGS PKGDVDPFYY DYE Highest antigen sequence identity to the following orthologs - mouse 46%, rat 63%.
The human FXYD2 (pronounced fix-id) gene maps to chromosome 11q23 and encodes the gamma subunit of Na/K-ATPase (NKA). FXYD2 gamma subunit modulates NKA activity by inducing ion channel activity. The mammalian FXYD family maintains Na+ and K+ gradients between the intracellular and extracellular milieus of cells in processes such as renal Na+-reabsorption, muscle contraction and neuronal excitability. FXYDs are single-span membrane proteins that share a 35 amino acid signature domain, beginning with the sequence PFXYD and containing 7 invariant and 6 conserved amino acids. Other members of the FXYD family include FXYD1 (PLM, phospholemman), FXYD3 (Mat8, mammary tumor protein 8), FXYD4 (CHIF) and FXYD5 (RIC).
Specifications
FXYD2 | |
Polyclonal | |
Unconjugated | |
Fxyd2 | |
ATP1C; Atp1g1; ATPase, Na+/K+ transporting, gamma 1 polypeptide; FXTD domain-containing ion transport regulator 2; FXYD domain containing ion transport regulator 2; FXYD domain-containing ion transport regulator 2; FXYD2; gamma subunit of sodium potassium ATPase; GNAKATP; HOMG2; Na(+)/K(+) ATPase subunit gamma; regulator 2; Sodium Potassium ATPase; sodium pump gamma chain; sodium/potassium-transporting ATPase gamma chain; sodium/potassium-transporting ATPase subunit gamma | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
486 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P54710 | |
Fxyd2 | |
Recombinant Human FXYD2. Recombinant protein control fragment (Product #RP-107318). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction