Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ FXYD2 Recombinant Protein

Catalog No. p-3719070
Click to view available options
Quantity:
10 μg
25 μg

Human FXYD2 full-length ORF with GST-tag at N-terminal

This gene encodes a member of a family of small membrane proteins that share a 35-amino acid signature sequence domain, beginning with the sequence PFXYD and containing 7 invariant and 6 highly conserved amino acids. The approved human gene nomenclature for the family is FXYD-domain containing ion transport regulator. Mouse FXYD5 has been termed RIC (Related to Ion Channel). FXYD2, also known as the gamma subunit of the Na,K-ATPase, regulates the properties of that enzyme. FXYD1 (phospholemman), FXYD2 (gamma), FXYD3 (MAT-8), FXYD4 (CHIF), and FXYD5 (RIC) have been shown to induce channel activity in experimental expression systems. Transmembrane topology has been established for two family members (FXYD1 and FXYD2), with the N-terminus extracellular and the C-terminus on the cytoplasmic side of the membrane. The Type III integral membrane protein encoded by this gene is the gamma subunit of the Na,K-ATPase present on the plasma membrane. Although the Na,K-ATPase does not depend on the gamma subunit to be functional, it is thought that the gamma subunit modulates the enzyme's activity by inducing ion channel activity. Mutations in this gene have been associated with renal hypomagnesaemia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.

  • Theoretical MW: 32.78kDa
  • Preparation Method: In vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 Fast Flow
  • Storage Buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Accession Number AAH05302.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 486
Molecular Weight (g/mol) 32.78
Name FXYD2 (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Source Wheat Germ (in vitro)
Immunogen MDRWYLGGSPKGDVDPFYYDYETVRNGGLIFAGLAFIVGLLILLSRRFRCGGNKKRRQINEDEP
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias ATP1G1/HOMG2/MGC12372
Common Name FXYD2
Gene Symbol FXYD2
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.