Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FXYD3 Monoclonal Antibody (OTI2F7), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA504594

Description
FXYD3 belongs to a small family of FXYD-domain containing regulators of Na+/K+ ATPases which share a 35-amino acid signature sequence domain, beginning with the sequence PFXYD, and containing 7 invariant and 6 highly conserved amino acids. This gene encodes a cell membrane protein that may regulate the function of ion-pumps and ion-channels. This gene may also play a role in tumor progression. Alternative splicing results in multiple transcript variants encoding distinct isoforms.Specifications
FXYD3 | |
Monoclonal | |
0.48 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q14802 | |
FXYD3 | |
Full length human recombit protein of human FXYD3 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI2F7 | |
Unconjugated | |
FXYD3 | |
MAT8, PLML | |
Mouse | |
Affinity Chromatography | |
RUO | |
5349 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction