Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FXYD5/Dysadherin Antibody (M029), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP323349
Description
FXYD5/Dysadherin Monoclonal antibody specifically detects FXYD5/Dysadherin in Human samples. It is validated for Western Blot, Immunofluorescence, ImmunoprecipitationSpecifications
FXYD5/Dysadherin | |
Monoclonal | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence, Immunoprecipitation | |
M029 | |
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:100 | |
DYSAD, dysadherin, FXYD domain containing ion transport regulator 5, FXYD domain-containing ion transport regulator 5, IWU1, KCT1, keratinocytes associated transmembrane protein 1, OIT2, PRO6241, RIC | |
Clone (M029) was generated from a proprietary antigen related to the native human FXYD5/Dysadherin protein expressed in A431 epidermoid carcinoma cell line. [UniProt# Q96DB9] | |
0.1 mL | |
Signal Transduction | |
53827 | |
Store at -20°C. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction