Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FXYD6 Monoclonal Antibody (2H1B7), Proteintech
SDP

Catalog No. 502334157 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-233-4157 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-233-4157 Supplier Proteintech Group Inc Supplier No. 680581IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FXYD6 Monoclonal antibody specifically detects FXYD6 in Chicken, Human, Mouse, Pig, Rabbit, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen FXYD6
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2H1B7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Accession No. Q9H0Q3
Gene Alias 0610030I18Rik; FXYD6; Php; Plp; UNQ521/PRO1056
Host Species Mouse
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103349230
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.